siRNA Details
| Gene Symbol: MRPS21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018997.3 | NP_061870.1 | 1, 2 | 601 | ||
| NM_031901.5 | NP_114107.1 | 2, 3 | 165 | |||
| GenBank mRNA | AB049955.1 | - | 2, 3 | 138 | ||
| AF182417.1 | - | 1, 2 | 110 | |||
| AK289467.1 | - | 2, 3 | 152 | |||
| BC004566.2 | - | 2, 3 | 134 | 3633098 | ||
| BC006554.2 | - | 1, 2 | 506 | 2819715 | ||
| BI755132.1 | - | 1, 2 | 558 | |||
| HY161691.1 | - | 2, 3 | 166 | |||
| U79258.1 | - | 1, 2 | 522 | |||

MRPS21
Genomic Map