siRNA Details
| Gene Symbol: ANKIB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019004.1 | NP_061877.1 | 9 | 1670 | ||
| XM_005250458.3 | XP_005250515.1 | 9 | 1688 | |||
| XM_011516334.1 | XP_011514636.1 | 8 | 1505 | |||
| XM_011516335.2 | XP_011514637.1 | 9 | 1252 | |||
| XM_017012359.1 | XP_016867848.1 | 9 | 1688 | |||
| XM_017012360.1 | XP_016867849.1 | 5 | 463 | |||
| GenBank mRNA | AB037807.1 | - | 9 | 1670 | ||
| AK025475.1 | - | 6 | 767 | |||
| BC031483.1 | - | 2 | 199 | |||
| BC063861.1 | - | 9 | 1646 | |||
| BC073893.1 | - | 3 | 262 | |||

ANKIB1
Genomic Map