siRNA Details
| Gene Symbol: ARMCX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009584.1 | NP_001009584.1 | 2 | 331 | ||
| NM_001184768.1 | NP_001171697.1 | 4 | 501 | |||
| NM_019007.3 | NP_061880.2 | 3 | 413 | |||
| XM_011530976.2 | XP_011529278.1 | 4 | 467 | |||
| GenBank mRNA | AK097018.1 | - | 1 | 805 | ||
| BC007677.2 | - | 3 | 413 | 3609980 | ||
| DA933518.1 | - | 4 | 498 | |||

ARMCX6
Genomic Map