siRNA Details
| Gene Symbol: DGCR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190326.1 | NP_001177255.1 | 4 | 1357 | ||
| NM_022720.6 | NP_073557.3 | 4 | 1357 | |||
| GenBank mRNA | AK025539.1 | - | 3 | 321 | ||
| AK222796.1 | - | 3 | 321 | |||
| BC009323.2 | - | 2 | 160 | |||
| BC037564.1 | - | 4 | 1307 | |||
| BC078147.1 | - | 3 | 1071 | 6014346 | ||
| BX649187.1 | - | 2 | 85 | |||
| CR456356.1 | - | 3 | 1015 | |||

DGCR8
Genomic Map