siRNA Details
| Gene Symbol: C11orf71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271562.1 | NP_001258491.1 | 1 | 516 | ||
| NM_019021.3 | NP_061894.2 | 1 | 516 | |||
| GenBank mRNA | AB101209.1 | - | 1 | 505 | ||
| AK000017.1 | - | 1 | 477 | |||
| BC009041.1 | - | 1 | 445 | 3868744 | ||
| BC071695.1 | - | 1 | 383 | 5416335 | ||
| CR457222.1 | - | 1 | 297 | |||

C11orf71
Genomic Map