siRNA Details
| Gene Symbol: TMX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019022.3 | NP_061895.3 | 6 | 470 | ||
| XM_005266714.2 | XP_005266771.1 | 5 | 415 | |||
| XM_011526034.2 | XP_011524336.1 | 6 | 462 | |||
| XM_011526035.2 | XP_011524337.1 | 6 | 462 | |||
| GenBank mRNA | AB058733.1 | - | 6 | 432 | ||
| AK122715.1 | - | 6 | 462 | |||
| AK307269.1 | - | 5 | 343 | |||
| AK310389.1 | - | 6 | 652 | |||
| AL832341.1 | - | 6 | 418 | |||
| AL832356.1 | - | 6 | 418 | |||
| BC007690.2 | - | 5 | 261 | |||
| BC018981.2 | - | 5 | 261 | |||
| BC032325.2 | - | 6 | 423 | |||
| BC093792.1 | - | 6 | 430 | 7939637 | ||
| BC093794.1 | - | 6 | 431 | 7939639 | ||
| BC107422.1 | - | 6 | 466 | |||
| BC143400.1 | - | 5 | 385 | |||
| BX647846.1 | - | 6 | 479 | |||

TMX3
Genomic Map