siRNA Details
| Gene Symbol: HEATR5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019024.2 | NP_061897.1 | 4 | 502 | ||
| XM_006712034.2 | XP_006712097.1 | 4 | 441 | |||
| XM_011532934.2 | XP_011531236.1 | 4 | 969 | |||
| XM_017004378.1 | XP_016859867.1 | 4 | 441 | |||
| XM_017004379.1 | XP_016859868.1 | 5 | 879 | |||
| XM_017004380.1 | XP_016859869.1 | 5 | 652 | |||
| GenBank mRNA | AK096637.1 | - | 4 | 441 | ||
| AV759566.1 | - | 4 | 498 | |||
| BC150605.1 | - | 4 | 487 | 9056975 | ||

HEATR5B
Genomic Map