siRNA Details
| Gene Symbol: CPVL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019029.2 | NP_061902.2 | 6, 7 | 746 | ||
| NM_031311.3 | NP_112601.3 | 6, 7 | 658 | |||
| XM_011515437.1 | XP_011513739.1 | 6, 7 | 825 | |||
| XM_017012366.1 | XP_016867855.1 | 6, 7 | 658 | |||
| GenBank mRNA | AF106704.2 | - | 6, 7 | 578 | ||
| AF217508.1 | - | 5, 6 | 545 | |||
| AF282617.1 | - | 6, 7 | 608 | |||
| AK075433.1 | - | 6, 7 | 659 | |||
| AK124472.1 | - | 10, 11 | 1186 | |||
| AK225545.1 | - | 6, 7 | 609 | |||
| AY358549.1 | - | 6, 7 | 635 | |||
| BC016838.2 | - | 6, 7 | 706 | 3888647 | ||

CPVL
Genomic Map