siRNA Details
| Gene Symbol: RHOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019034.2 | NP_061907.2 | 3, 4 | 392 | ||
| GenBank mRNA | AF239923.1 | - | 3, 4 | 366 | ||
| AK000254.1 | - | 3, 4 | 358 | |||
| AK289482.1 | - | 3, 4 | 966 | |||
| BC018208.1 | - | 3, 4 | 371 | 3864426 | ||
| BQ053707.1 | - | 3, 4 | 373 | |||
| BU527972.1 | - | 3, 4 | 374 | |||

RHOF
Genomic Map