siRNA Details
| Gene Symbol: PUS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318163.1 | NP_001305092.1 | 1 | 143 | ||
| NM_001318164.1 | NP_001305093.1 | 2 | 350 | |||
| NM_019042.4 | NP_061915.2 | 2 | 374 | |||
| XM_005250462.3 | XP_005250519.1 | 2 | 382 | |||
| XM_011516336.2 | XP_011514638.1 | 2 | 377 | |||
| XM_017012367.1 | XP_016867856.1 | 2 | 310 | |||
| XM_017012368.1 | XP_016867857.1 | 2 | 386 | |||
| GenBank mRNA | AB067484.1 | - | 1 | 70 | ||
| AK091283.1 | - | 2 | 354 | |||
| AK128629.1 | - | 2 | 336 | |||
| BC011396.1 | - | 2 | 330 | 3870711 | ||
| CN481800.1 | - | 2 | 350 | |||

PUS7
Genomic Map