siRNA Details
| Gene Symbol: CCDC93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019044.4 | NP_061917.3 | 17, 18 | 1513 | ||
| XM_011511359.1 | XP_011509661.1 | 17, 18 | 1510 | |||
| XM_011511361.1 | XP_011509663.1 | 16, 17 | 1267 | |||
| XM_011511362.1 | XP_011509664.1 | 16, 17 | 1381 | |||
| GenBank mRNA | AB209993.1 | - | 17, 18 | 1432 | ||
| AK001858.1 | - | 5, 6 | 307 | |||
| AK025611.1 | - | 16, 17 | 1281 | |||
| AK057926.1 | - | 17, 18 | 1374 | |||
| AK290722.1 | - | 17, 18 | 1420 | |||
| AK309040.1 | - | 17, 18 | 1420 | |||
| BC028609.1 | - | 17, 18 | 1486 | 4826022 | ||
| BC093018.1 | - | 17, 18 | 1403 | 30337330 | ||

CCDC93
Genomic Map