siRNA Details
| Gene Symbol: ANKRD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009941.2 | NP_001009941.1 | 4 | 1136 | ||
| NM_001009943.2 | NP_001009943.1 | 4 | 1136 | |||
| NM_019046.2 | NP_061919.1 | 4 | 1136 | |||
| GenBank mRNA | AW630443.1 | - | 3 | 215 | ||
| BC062346.1 | - | 4 | 622 | 3845142 | ||

ANKRD16
Genomic Map