siRNA Details
| Gene Symbol: MIER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017550.1 | NP_060020.1 | 7, 8 | 668 | ||
| XM_005259582.4 | XP_005259639.1 | 7, 8 | 1252 | |||
| XM_006722769.3 | XP_006722832.1 | 7, 8 | 1256 | |||
| XM_011528072.2 | XP_011526374.1 | 7, 8 | 1254 | |||
| XM_011528073.1 | XP_011526375.1 | 7, 8 | 670 | |||
| XM_011528074.2 | XP_011526376.1 | 7, 8 | 1258 | |||
| XM_011528075.2 | XP_011526377.1 | 7, 8 | 1775 | |||
| XM_011528077.2 | XP_011526379.1 | 8, 9 | 3616 | |||
| XM_011528081.2 | XP_011526383.1 | 5, 6 | 512 | |||
| XM_011528082.2 | XP_011526384.1 | 5, 6 | 443 | |||
| GenBank mRNA | AB033019.1 | - | 656 | |||
| BC028203.1 | - | 7, 8 | 668 | 5201526 | ||

MIER2
Genomic Map