siRNA Details
| Gene Symbol: RC3H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100588.1 | NP_001094058.1 | 6 | 1050 | ||
| NM_018835.2 | NP_061323.2 | 6 | 1050 | |||
| GenBank mRNA | AF255303.1 | - | 5 | 1410 | ||
| AK000308.1 | - | 4 | 484 | |||
| AK226010.1 | - | 4 | 484 | |||
| AL833177.1 | - | 6 | 989 | |||
| BC044642.1 | - | 6 | 982 | |||

RC3H2
Genomic Map