siRNA Details
| Gene Symbol: TOMM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019059.3 | NP_061932.1 | 3 | 694 | ||
| GenBank mRNA | BC071973.1 | - | 3 | 644 | 6278846 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TOMM7| Gene Symbol: TOMM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019059.3 | NP_061932.1 | 3 | 694 | ||
| GenBank mRNA | BC071973.1 | - | 3 | 644 | 6278846 | |