siRNA Details
| Gene Symbol: DLL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019074.3 | NP_061947.1 | 2, 3 | 681 | ||
| GenBank mRNA | AB036931.1 | - | 2, 3 | 621 | ||
| AB043894.1 | - | 2, 3 | 345 | |||
| AF253468.1 | - | 2, 3 | 345 | |||
| AF279305.1 | - | 1, 2 | 251 | |||
| AK313831.1 | - | 2, 3 | 676 | |||
| AY358894.1 | - | 2, 3 | 380 | |||
| BC106950.2 | - | 2, 3 | 481 | 40034887 | ||

DLL4
Genomic Map