siRNA Details
| Gene Symbol: UGT1A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019076.4 | NP_061949.3 | 1 | 856 | ||
| GenBank mRNA | AF030310.1 | - | 1 | 793 | ||
| AF462267.1 | - | 1 | 826 | |||
| AF462268.1 | - | 1 | 856 | |||
| AK313488.1 | - | 1 | 839 | |||
| AY435143.1 | - | 1 | 793 | |||
| DQ364251.1 | - | 1 | 448 | |||
| Gene Symbol: UGT1A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019075.2 | NP_061948.1 | 1 | 839 | ||
| GenBank mRNA | AK025403.1 | - | 1 | 446 | ||
| AY435137.1 | - | 1 | 793 | |||
| BC020971.1 | - | 1 | 862 | 3850654 | ||
| BC053576.1 | - | 1 | 814 | 4452440 | ||
| BC069210.1 | - | 1 | 823 | 4660540 | ||
| U89508.1 | - | 1 | 793 | |||
| Gene Symbol: UGT1A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019077.2 | NP_061950.2 | 1 | 793 | ||
| GenBank mRNA | AY435142.1 | - | 1 | 793 | ||
| BC001521.2 | - | 1 | 363 | |||
| DQ383513.1 | - | 1 | 505 | |||
| U89507.1 | - | 1 | 793 | |||
| Gene Symbol: UGT1A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021027.2 | NP_066307.1 | 1 | 830 | ||
| GenBank mRNA | AF056188.1 | - | 1 | 811 | ||
| AK313510.1 | - | 1 | 869 | |||
| AY435144.1 | - | 1 | 793 | |||
| BC058844.1 | - | 1 | 836 | 4593613 | ||
| DQ364246.1 | - | 1 | 145 | |||
| S55985.1 | - | 1 | 827 | |||

UGT1A8
Genomic Map