siRNA Details
| Gene Symbol: LZTFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276378.1 | NP_001263307.1 | 10 | 1113 | ||
| NM_001276379.1 | NP_001263308.1 | 9 | 988 | |||
| NM_020347.3 | NP_065080.1 | 8 | 832 | |||
| NR_075080.1 | 7 | 707 | ||||
| XM_006713207.2 | XP_006713270.2 | 8 | 817 | |||
| XM_011533838.2 | XP_011532140.1 | 8 | 756 | |||
| XM_017006644.1 | XP_016862133.1 | 11 | 1470 | |||
| XM_017006645.1 | XP_016862134.1 | 10 | 1345 | |||
| GenBank mRNA | AJ297351.1 | - | 8 | 789 | ||
| AK093705.1 | - | 10 | 1431 | |||
| BC025988.2 | - | 8 | 761 | 4838035 | ||
| BC042483.1 | - | 8 | 792 | 4831113 | ||
| BX640604.1 | - | 8 | 817 | |||
| CN429018.1 | - | 7 | 617 | |||

LZTFL1
Genomic Map