siRNA Details
| Gene Symbol: EQTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161585.1 | NP_001155057.1 | 7 | 659 | ||
| NM_020641.2 | NP_065692.2 | 8 | 746 | |||
| XM_011517920.2 | XP_011516222.1 | 5 | 1425 | |||
| GenBank mRNA | AJ278482.1 | - | 8 | 746 | ||
| BC137351.1 | - | 8 | 716 | 9021349 | ||
| BC137352.1 | - | 8 | 716 | 9021350 | ||
| BC144602.1 | - | 7 | 629 | 9053133 | ||
| BX089774.1 | - | 2 | 131 | |||

EQTN
Genomic Map