siRNA Details
| Gene Symbol: EQTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161585.1 | NP_001155057.1 | 1, 2 | 160 | ||
| NM_020641.2 | NP_065692.2 | 1, 2 | 160 | |||
| GenBank mRNA | AJ278482.1 | - | 1, 2 | 160 | ||
| BC014307.1 | - | 1, 2 | 160 | |||
| BC137351.1 | - | 1, 2 | 130 | 9021349 | ||
| BC137352.1 | - | 1, 2 | 130 | 9021350 | ||
| BC144602.1 | - | 1, 2 | 130 | 9053133 | ||

EQTN
Genomic Map