siRNA Details
| Gene Symbol: UGT1A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007120.2 | NP_009051.1 | 1 | 295 | ||
| GenBank mRNA | AK313623.1 | - | 1 | 309 | ||
| AY435139.1 | - | 1 | 266 | |||
| BC121036.1 | - | 1 | 266 | 40117585 | ||
| BC131623.1 | - | 1 | 259 | |||
| BC139784.1 | - | 1 | 286 | 40117089 | ||
| DQ364249.1 | - | 1 | 275 | |||
| M57951.1 | - | 1 | 295 | |||

UGT1A4
Genomic Map