siRNA Details
| Gene Symbol: TMEM106B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134232.1 | NP_001127704.1 | 8 | 946 | ||
| NM_018374.3 | NP_060844.2 | 9 | 1140 | |||
| XM_005249789.1 | XP_005249846.1 | 8 | 996 | |||
| GenBank mRNA | AK001834.1 | - | 1 | 443 | ||
| AK002135.1 | - | 8 | 870 | |||
| AK223263.1 | - | 8 | 844 | |||
| BC028108.1 | - | 8 | 860 | |||
| BC033901.2 | - | 9 | 1062 | 5263860 | ||
| BC039741.2 | - | 8 | 857 | 6066301 | ||

TMEM106B
Genomic Map