siRNA Details
| Gene Symbol: LRRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099658.1 | NP_001093128.1 | 3 | 1904 | ||
| NM_001099660.1 | NP_001093130.1 | 4 | 2011 | |||
| NM_018334.4 | NP_060804.3 | 2 | 1822 | |||
| GenBank mRNA | AB060967.1 | - | 1 | 1271 | ||
| AF134481.1 | - | 1 | 1302 | |||
| AK292556.1 | - | 2 | 1411 | |||
| AL442092.1 | - | 1 | 1227 | |||
| AY358280.1 | - | 3 | 1444 | |||
| BC035133.2 | - | 2 | 1701 | 5263900 | ||
| CR457397.1 | - | 1 | 965 | |||

LRRN3
Genomic Map