siRNA Details
| Gene Symbol: ZNHIT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170670.1 | NP_001164141.1 | 2, 3 | 697 | ||
| NM_017953.3 | NP_060423.3 | 1, 2 | 814 | |||
| XM_011541614.2 | XP_011539916.1 | 1, 2 | 828 | |||
| GenBank mRNA | AF308296.1 | - | 1, 2 | 461 | ||
| AK000736.1 | - | 1, 2 | 186 | |||
| AK000767.1 | - | 1, 2 | 723 | |||
| AK298149.1 | - | 2, 3 | 697 | |||
| AK314566.1 | - | 1, 2 | 742 | |||
| AL442074.1 | - | 1, 2 | 799 | |||
| BC026236.1 | - | 1, 2 | 801 | 4664103 | ||
| BC110898.1 | - | 1, 2 | 732 | 6012059 | ||

ZNHIT6
Genomic Map