siRNA Details
| Gene Symbol: GPN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018066.3 | NP_060536.3 | 3 | 969 | ||
| GenBank mRNA | AK001211.1 | - | 3 | 888 | ||
| AK022535.1 | - | 3 | 874 | |||
| AY358864.1 | - | 3 | 878 | |||
| AY364261.1 | - | 3 | 687 | |||
| BC007815.2 | - | 3 | 811 | 4299794 | ||
| BC020272.1 | - | 3 | 854 | |||

GPN2
Genomic Map