siRNA Details
| Gene Symbol: RAB39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017516.2 | NP_059986.1 | 2 | 544 | ||
| GenBank mRNA | AK292969.1 | - | 2 | 516 | ||
| BC028064.1 | - | 2 | 468 | 5197104 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB39A| Gene Symbol: RAB39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017516.2 | NP_059986.1 | 2 | 544 | ||
| GenBank mRNA | AK292969.1 | - | 2 | 516 | ||
| BC028064.1 | - | 2 | 468 | 5197104 | ||