siRNA Details
| Gene Symbol: RAB39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017516.2 | NP_059986.1 | 1, 2 | 318 | ||
| GenBank mRNA | AK292969.1 | - | 1, 2 | 290 | ||
| BC028064.1 | - | 1, 2 | 242 | 5197104 | ||
| HY107079.1 | - | 1, 2 | 318 | |||
| X99962.1 | - | 1, 2 | 224 | |||

RAB39A
Genomic Map