siRNA Details
| Gene Symbol: EPDR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242948.1 | NP_001229877.1 | 2 | 333 | ||
| NM_017549.4 | NP_060019.2 | 2 | 848 | |||
| GenBank mRNA | AJ250475.2 | - | 2 | 321 | ||
| AK290885.1 | - | 2 | 542 | |||
| AK309180.1 | - | 2 | 320 | |||
| AY027862.2 | - | 2 | 848 | |||
| BC000686.2 | - | 2 | 592 | |||
| BC018299.1 | - | 2 | 740 | 3461888 | ||
| DA938402.1 | - | 3 | 361 | |||
| DB167672.1 | - | 2 | 334 | |||

EPDR1
Genomic Map