siRNA Details
| Gene Symbol: KLHDC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184854.1 | NP_001171783.1 | 2 | 230 | ||
| NM_001184856.1 | NP_001171785.1 | 4 | 401 | |||
| NM_017566.3 | NP_060036.2 | 4 | 401 | |||
| XM_005255994.3 | XP_005256051.1 | 4 | 406 | |||
| XM_005255999.1 | XP_005256056.1 | 3 | 321 | |||
| XM_006721201.1 | XP_006721264.1 | 4 | 406 | |||
| XM_006721202.1 | XP_006721265.1 | 4 | 406 | |||
| XM_006721203.2 | XP_006721266.1 | 4 | 406 | |||
| XM_006721204.3 | XP_006721267.1 | 4 | 406 | |||
| XM_017023344.1 | XP_016878833.1 | 4 | 406 | |||
| XM_017023345.1 | XP_016878834.1 | 4 | 346 | |||
| GenBank mRNA | AK023986.1 | - | 4 | 358 | ||
| AK097097.1 | - | 2 | 1282 | |||
| BC001044.2 | - | 2 | 137 | 3505126 | ||
| BC022969.1 | - | 4 | 345 | 5112326 | ||
| BP215584.1 | - | 4 | 401 | |||

KLHDC4
Genomic Map