siRNA Details
| Gene Symbol: ROPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317774.1 | NP_001304703.1 | 6 | 964 | ||
| NM_001317775.1 | NP_001304704.1 | 5 | 836 | |||
| NM_017578.3 | NP_060048.2 | 7 | 1044 | |||
| XM_011512933.2 | XP_011511235.1 | 7 | 1008 | |||
| XM_011512934.2 | XP_011511236.1 | 7 | 1119 | |||
| GenBank mRNA | AF303889.2 | - | 7 | 1044 | ||
| BC132744.1 | - | 5 | 800 | 40146766 | ||
| BC132746.1 | - | 5 | 801 | 40146768 | ||
| BG219480.1 | - | 2 | 312 | |||

ROPN1
Genomic Map