siRNA Details
| Gene Symbol: RNF111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270528.1 | NP_001257457.1 | 6 | 1815 | ||
| NM_001270529.1 | NP_001257458.1 | 6 | 1815 | |||
| NM_001270530.1 | NP_001257459.1 | 6 | 1672 | |||
| NM_017610.7 | NP_060080.6 | 6 | 1815 | |||
| XM_005254479.3 | XP_005254536.1 | 6 | 1855 | |||
| XM_006720575.3 | XP_006720638.1 | 7 | 2026 | |||
| XM_006720579.3 | XP_006720642.1 | 6 | 1672 | |||
| XM_011521694.2 | XP_011519996.2 | 6 | 1808 | |||
| XM_011521696.2 | XP_011519998.2 | 6 | 1502 | |||
| XM_011521697.2 | XP_011519999.2 | 6 | 1502 | |||
| XM_011521699.2 | XP_011520001.2 | 6 | 1497 | |||
| XM_017022336.1 | XP_016877825.1 | 7 | 1945 | |||
| XM_017022337.1 | XP_016877826.1 | 6 | 1855 | |||
| XM_017022338.1 | XP_016877827.1 | 6 | 1893 | |||
| XM_017022339.1 | XP_016877828.1 | 6 | 1855 | |||
| XM_017022340.1 | XP_016877829.1 | 6 | 1855 | |||
| XM_017022341.1 | XP_016877830.1 | 6 | 1672 | |||
| XM_017022342.1 | XP_016877831.1 | 6 | 1669 | |||
| XM_017022343.1 | XP_016877832.1 | 6 | 1672 | |||
| GenBank mRNA | AK131304.1 | - | 6 | 1673 | ||
| AK131488.1 | - | 5 | 675 | |||
| AL832089.1 | - | 3 | 400 | |||
| BC060862.1 | - | 6 | 1818 | 30336202 | ||
| BX538130.1 | - | 6 | 1816 | |||
| BX647259.1 | - | 6 | 1529 | |||

RNF111
Genomic Map