siRNA Details
| Gene Symbol: BORCS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017622.2 | NP_060092.2 | 1 | 1043 | ||
| GenBank mRNA | AK000021.1 | - | 1 | 869 | ||
| AK222506.1 | - | 1 | 869 | |||
| BC009261.2 | - | 1 | 826 | 3928189 | ||
| BC018880.2 | - | 1 | 925 | 3615792 | ||
| BC070272.1 | - | 1 | 940 | 4672989 | ||

BORCS6
Genomic Map