siRNA Details
| Gene Symbol: BORCS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017622.2 | NP_060092.2 | 1 | 1036 | ||
| GenBank mRNA | AK000021.1 | - | 1 | 862 | ||
| AK222506.1 | - | 1 | 862 | |||
| BC009261.2 | - | 1 | 819 | 3928189 | ||
| BC018880.2 | - | 1 | 918 | 3615792 | ||
| BC070272.1 | - | 1 | 933 | 4672989 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_028005, AK009886, AK169875, AK151629.

BORCS6
Genomic Map