siRNA Details
| Gene Symbol: KCTD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017634.3 | NP_060104.2 | 1, 2 | 278 | ||
| GenBank mRNA | AK298518.1 | - | 1, 2 | 82 | ||
| BC021216.2 | - | 1, 2 | 81 | 4302317 | ||
| BC068518.1 | - | 1, 2 | 91 | 5265435 | ||
| BG249272.1 | - | 217 | ||||
| BU149129.1 | - | 1, 2 | 196 | |||
| DA115324.1 | - | 1, 2 | 279 | |||

KCTD9
Genomic Map