siRNA Details
| Gene Symbol: KCTD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017634.3 | NP_060104.2 | 5 | 579 | ||
| GenBank mRNA | AK298518.1 | - | 5 | 383 | ||
| BC021216.2 | - | 5 | 382 | 4302317 | ||
| BC068518.1 | - | 5 | 392 | 5265435 | ||
| BG249272.1 | - | 513 | ||||
| BU149129.1 | - | 5 | 497 | |||

KCTD9
Genomic Map