siRNA Details
| Gene Symbol: BNC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317939.1 | NP_001304868.1 | 5 | 1530 | ||
| NM_001317940.1 | NP_001304869.1 | 5 | 1436 | |||
| NM_017637.5 | NP_060107.3 | 6 | 1656 | |||
| XM_011517924.1 | XP_011516226.1 | 5 | 1580 | |||
| XM_011517934.2 | XP_011516236.1 | 3 | 2608 | |||
| XM_017014816.1 | XP_016870305.1 | 8 | 3966 | |||
| XM_017014817.1 | XP_016870306.1 | 7 | 2115 | |||
| XM_017014818.1 | XP_016870307.1 | 6 | 2030 | |||
| XM_017014819.1 | XP_016870308.1 | 7 | 2114 | |||
| XM_017014820.1 | XP_016870309.1 | 7 | 2114 | |||
| XM_017014821.1 | XP_016870310.1 | 6 | 2028 | |||
| XM_017014822.1 | XP_016870311.1 | 4 | 1576 | |||
| XM_017014823.1 | XP_016870312.1 | 4 | 6345 | |||
| XM_017014824.1 | XP_016870313.1 | 3 | 1897 | |||
| XM_017014825.1 | XP_016870314.1 | 7 | 2113 | |||
| XM_017014826.1 | XP_016870315.1 | 7 | 2113 | |||
| XM_017014827.1 | XP_016870316.1 | 7 | 2113 | |||
| XM_017014828.1 | XP_016870317.1 | 7 | 2112 | |||
| XM_017014829.1 | XP_016870318.1 | 5 | 2084 | |||
| GenBank mRNA | AK092247.1 | - | 1 | 386 | ||
| AK131398.1 | - | 6 | 1538 | |||
| AK299073.1 | - | 5 | 1406 | |||
| AK304745.1 | - | 3 | 1198 | |||
| AY438376.1 | - | 4 | 1301 | |||
| BX641030.1 | - | 1 | 687 | |||
| CR933649.1 | - | 5 | 1487 | |||

BNC2
Genomic Map