siRNA Details
| Gene Symbol: AFTPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002243.2 | NP_001002243.1 | 6 | 2699 | ||
| NM_017657.4 | NP_060127.3 | 6 | 2699 | |||
| NM_203437.3 | NP_982261.2 | 6 | 2699 | |||
| XM_005264380.1 | XP_005264437.1 | 6 | 2699 | |||
| GenBank mRNA | AK000087.1 | - | 6 | 2674 | ||
| AK021899.1 | - | 5 | 1322 | |||
| AK024658.1 | - | 4 | 335 | |||
| AK222821.1 | - | 6 | 2622 | |||
| AL833962.2 | - | 6 | 2477 | |||
| AY367088.1 | - | 5 | 2359 | |||
| BC022247.1 | - | 3 | 191 | |||
| BC047529.1 | - | 6 | 2673 | 5311682 | ||

AFTPH
Genomic Map