siRNA Details
| Gene Symbol: QPCTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163377.1 | NP_001156849.1 | 3 | 603 | ||
| NM_017659.3 | NP_060129.2 | 4 | 885 | |||
| XM_011527048.2 | XP_011525350.1 | 4 | 967 | |||
| XM_017026900.1 | XP_016882389.1 | 4 | 964 | |||
| GenBank mRNA | AK000091.1 | - | 4 | 694 | ||
| AK172764.1 | - | 4 | 693 | |||
| AK222636.1 | - | 4 | 678 | |||
| BC011553.1 | - | 3 | 389 | 4564051 | ||

QPCTL
Genomic Map