siRNA Details
| Gene Symbol: BEST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017682.2 | NP_060152.2 | 5 | 686 | ||
| XM_005259963.3 | XP_005260020.1 | 6 | 886 | |||
| GenBank mRNA | AF440756.1 | - | 5 | 686 | ||
| AK000139.1 | - | 5 | 621 | |||
| AK225232.1 | - | 5 | 621 | |||
| AY515705.1 | - | 5 | 686 | |||

BEST2
Genomic Map