siRNA Details
| Gene Symbol: GDAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135589.2 | NP_001129061.1 | 2 | 421 | ||
| NM_017686.3 | NP_060156.1 | 2 | 421 | |||
| XM_005270959.2 | XP_005271016.1 | 2 | 421 | |||
| XM_011541621.2 | XP_011539923.1 | 2 | 421 | |||
| XM_011541622.1 | XP_011539924.1 | 2 | 309 | |||
| XM_011541623.2 | XP_011539925.1 | 2 | 421 | |||
| XM_011541624.2 | XP_011539926.1 | 2 | 784 | |||
| XM_017001531.1 | XP_016857020.1 | 2 | 363 | |||
| XM_017001532.1 | XP_016857021.1 | 2 | 784 | |||
| GenBank mRNA | AK000149.1 | - | 2 | 324 | ||
| BC013132.1 | - | 2 | 304 | 3448678 | ||
| BJ992034.1 | - | 2 | 344 | |||
| DB043931.1 | - | 2 | 424 | |||

GDAP2
Genomic Map