siRNA Details
| Gene Symbol: ARHGEF38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242729.1 | NP_001229658.1 | 1 | 324 | ||
| NM_017700.1 | NP_060170.1 | 1 | 324 | |||
| XM_011532050.2 | XP_011530352.1 | 1 | 61 | |||
| XM_011532051.2 | XP_011530353.1 | 1 | 339 | |||
| GenBank mRNA | AK000191.1 | - | 1 | 324 | ||
| BC105695.1 | - | 1 | 312 | 40023595 | ||

ARHGEF38
Genomic Map