siRNA Details
| Gene Symbol: FAM46C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017709.3 | NP_060179.2 | 2 | 293 | ||
| GenBank mRNA | AK000209.1 | - | 2 | 228 | ||
| BC036516.2 | - | 2 | 222 | 5267660 | ||
| BC131726.1 | - | 2 | 206 | 40123993 | ||
| CD300120.1 | - | 2 | 319 | |||

FAM46C
Genomic Map