siRNA Details
| Gene Symbol: ELP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031703.2 | NP_001026873.2 | 7 | 850 | ||
| XM_005265241.3 | XP_005265298.1 | 6 | 776 | |||
| XM_017006669.1 | XP_016862158.1 | 7 | 876 | |||
| XM_017006670.1 | XP_016862159.1 | 7 | 1121 | |||
| GenBank mRNA | AK225721.1 | - | 7 | 832 | ||
| AK298202.1 | - | 6 | 785 | |||
| BC000623.2 | - | 7 | 754 | 3346325 | ||
| BC014204.2 | - | 7 | 754 | 4620013 | ||
| DQ787088.1 | - | 7 | 731 | |||

ELP6
Genomic Map