siRNA Details
| Gene Symbol: CEACAM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001817.3 | NP_001808.2 | 7 | 799 | ||
| XM_006722977.3 | XP_006723040.1 | 6 | 694 | |||
| XM_011526348.2 | XP_011524650.1 | 6 | 1073 | |||
| XM_011526357.2 | XP_011524659.1 | 6 | 452 | |||
| XM_017026205.1 | XP_016881694.1 | 7 | 853 | |||
| XM_017026207.1 | XP_016881696.1 | 6 | 748 | |||
| GenBank mRNA | AB924370.1 | - | 6 | 570 | ||
| BF819374.1 | - | 6 | 372 | |||
| D90276.1 | - | 7 | 785 | |||

CEACAM4
Genomic Map