siRNA Details
| Gene Symbol: MS4A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017716.2 | NP_060186.2 | 3 | 368 | ||
| XM_011545117.2 | XP_011543419.1 | 4 | 516 | |||
| GenBank mRNA | AK000224.1 | - | 3 | 368 | ||
| BC029793.1 | - | 3 | 356 | 5185424 | ||
| BP262076.1 | - | 3 | 370 | |||

MS4A12
Genomic Map