siRNA Details
| Gene Symbol: TMEM214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083590.1 | NP_001077059.1 | 3 | 559 | ||
| NM_017727.4 | NP_060197.4 | 3 | 559 | |||
| XM_005264381.3 | XP_005264438.1 | 3 | 532 | |||
| XM_005264382.3 | XP_005264439.1 | 3 | 532 | |||
| GenBank mRNA | AK074981.1 | - | 3 | 487 | ||
| AK097308.1 | - | 3 | 489 | |||
| AK297150.1 | - | 3 | 500 | |||
| AK315359.1 | - | 1 | 23 | |||
| AL833725.1 | - | 3 | 419 | |||
| BC068479.1 | - | 3 | 511 | 4814193 | ||

TMEM214
Genomic Map