siRNA Details
| Gene Symbol: FNBP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024948.2 | NP_001020119.1 | 9 | 1049 | ||
| NM_001164473.2 | NP_001157945.1 | 9 | 1049 | |||
| NM_017737.4 | NP_060207.2 | 9 | 1049 | |||
| XM_011541625.2 | XP_011539927.1 | 9 | 1113 | |||
| XM_017001533.1 | XP_016857022.1 | 10 | 1244 | |||
| GenBank mRNA | AK000282.1 | - | 3 | 301 | ||
| AK299760.1 | - | 4 | 435 | |||
| AK299943.1 | - | 3 | 345 | |||
| AK315981.1 | - | 1 | 59 | |||
| AY514449.1 | - | 9 | 898 | |||
| BC062477.1 | - | 4 | 500 | |||
| BC074891.2 | - | 3 | 279 | |||
| BC074892.2 | - | 3 | 279 | |||

FNBP1L
Genomic Map