siRNA Details
| Gene Symbol: TEX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161584.1 | NP_001155056.1 | 4 | 1201 | ||
| NM_017746.3 | NP_060216.2 | 4 | 1257 | |||
| XM_011518798.1 | XP_011517100.1 | 4 | 1219 | |||
| XM_017014848.1 | XP_016870337.1 | 4 | 746 | |||
| GenBank mRNA | AB060968.1 | - | 4 | 1173 | ||
| AK000294.1 | - | 4 | 1173 | |||
| AK298916.1 | - | 6 | 780 | |||
| AK302619.1 | - | 4 | 1204 | |||
| AK303662.1 | - | 4 | 697 | |||
| AK304869.1 | - | 3 | 974 | |||
| AY598337.1 | - | 3 | 1042 | |||
| BC030652.2 | - | 4 | 1093 | 5268105 | ||

TEX10
Genomic Map