siRNA Details
| Gene Symbol: CWC25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017748.4 | NP_060218.1 | 9 | 1187 | ||
| NR_073428.1 | 10 | 1304 | ||||
| GenBank mRNA | AK000298.1 | - | 9 | 1137 | ||
| AK225373.1 | - | 9 | 1157 | |||
| AK296115.1 | - | 10 | 1269 | |||
| AK316077.1 | - | 8 | 1133 | |||
| BC008833.2 | - | 9 | 1173 | 4304656 | ||
| CR749362.1 | - | 5 | 369 | |||

CWC25
Genomic Map