siRNA Details
Gene Symbol: UHRF1BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017754.3 | NP_060224.3 | 15 | 3542 | ||
GenBank mRNA | AB126777.1 | - | 15 | 3377 | ||
AK127691.1 | - | 2 | 338 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: UHRF1BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017754.3 | NP_060224.3 | 15 | 3542 | ||
GenBank mRNA | AB126777.1 | - | 15 | 3377 | ||
AK127691.1 | - | 2 | 338 |